Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human P-Selectin/CD62P Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BBA32
This item is not returnable.
View return policy
Description
P-Selectin/CD62P Polyclonal specifically detects P-Selectin/CD62P in Human samples. It is validated for Western Blot, Adhesion Blockade.Specifications
P-Selectin/CD62P | |
Polyclonal | |
Unconjugated | |
CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP | |
Sheep | |
Protein A or G purified | |
RUO | |
6403 | |
Reconstitute with sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Western Blot, Adhesion Blockade | |
LYOPH | |
Western Blot 1 ug/mL, Adhesion Blockade | |
SELP | |
Chinese hamster ovary cell line CHO-derived recombinant human P-Selectin | |
500 μg | |
Primary | |
Selected for its ability to inhibit the binding of U937 cells to immobilized rhP-Selectin. In Western blots, less than 1% cross-reactivity is observed with rhE-Selectin and rhL-Selectin. | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction