Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human P-Selectin/CD62P Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Supplier: R&D Systems BBA34
This item is not returnable.
View return policy
Description
P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Human samples. It is validated for Flow Cytometry.Specifications
P-Selectin/CD62P | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
SELP | |
Recombinant human P-Selectin Extracellular domain | |
100 Tests | |
Primary | |
This antibody binds to CHO cells transfected with human P-Selectin but not to CHO cells transfected with either human E-Selectin or human L-Selectin. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
90 | |
Flow Cytometry 10 uL/10^6 cells | |
CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6403 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction