Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human p21/CIP1/CDKN1A Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC1047U100UG
This item is not returnable.
View return policy
Description
p21/CIP1/CDKN1A Monoclonal specifically detects p21/CIP1/CDKN1A in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
p21/CIP1/CDKN1A | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P38936 | |
CDKN1A | |
E. coli-derived recombinant human p21/CIP1/CDKN1A Ser2-Pro164 Accession # P38936 | |
RUO | |
1026 | |
Human | |
IgG2a |
Flow Cytometry | |
195720 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1A, CDKN1melanoma differentiation associated protein 6, CIP1, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1 | |
Mouse | |
100 μg | |
Primary | |
Detects human p21/CIP1/CDKN1A in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction