Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human P2X7/P2RX7 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$188.28 - $532.27

Specifications

Antigen P2X7/P2RX7
Clone 545230
Dilution Immunocytochemistry 5-25 ug/mL
Applications Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB9790SP
GREENER_CHOICE
View Documents
R&D Systems
MAB9790SP
25 μg
Each for $188.28
Only null left
Add to Cart
 
MAB9790100
GREENER_CHOICE
View Documents
R&D Systems
MAB9790100
100 μg
Each for $532.27
Only null left
Add to Cart
 
Description

Description

P2X7/P2RX7 Monoclonal specifically detects P2X7/P2RX7 in Human samples. It is validated for Immunocytochemistry.
Specifications

Specifications

P2X7/P2RX7
Immunocytochemistry 5-25 ug/mL
Monoclonal
Supernatant
Human
Q99572
5027
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
545230
Immunocytochemistry
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
ATP receptor, MGC20089, P2RX7, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A
E. coli-derived recombinant human P2X7/P2RX7 Asp356-Tyr595 Accession # Q99572
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human P2X7/P2RX7 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.