Learn More
Human P2Y1/P2RY1 PE-conjugated Antibody, R&D Systems™
Description
Specifications
Specifications
| Antigen | P2Y1/P2RY1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 479509 |
| Conjugate | PE |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | NP_002554 |
| Gene Alias | ATP receptor, P2 purinoceptor subtype Y1, P2RY1, P2Y purinoceptor 1, P2Y1platelet ADP receptor, Purinergic receptor, purinergic receptor P2Y, G-protein coupled, 1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.