Learn More
Human P2Y10/P2RY10 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | P2Y10/P2RY10 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 639329 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | O00398 |
| Gene Alias | G-protein coupled purinergic receptor P2Y10, P2RY10, P2Y purinoceptor 10, P2Y10P2Y-like receptor, purinergic receptor P2Y, G-protein coupled, 10, putative P2Y purinoceptor 10 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.