Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human P2Y10/P2RY10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $456.00
Specifications
Antigen | P2Y10/P2RY10 |
---|---|
Clone | 639329 |
Dilution | Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 8-25 μg/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10158SP
![]() |
R&D Systems
MAB10158SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10158100
![]() |
R&D Systems
MAB10158100 |
100 μg |
Each for $456.00
|
|
|||||
Description
P2Y10/P2RY10 Monoclonal specifically detects P2Y10/P2RY10 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
P2Y10/P2RY10 | |
Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 8-25 μg/mL, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Human | |
O00398 | |
27334 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
639329 | |
Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
G-protein coupled purinergic receptor P2Y10, P2RY10, P2Y purinoceptor 10, P2Y10P2Y-like receptor, purinergic receptor P2Y, G-protein coupled, 10, putative P2Y purinoceptor 10 | |
NS0 mouse myeloma cell line transfected with human P2RY10, Accession # O00398 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human P2RY10 in Flow Cytometry and Immunohistochemistry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title