Learn More
Human P2Y11/P2RY11 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Description
Specifications
Specifications
| Antigen | P2Y11/P2RY11 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 505214 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | Q96G91 |
| Gene Alias | P2RY11, P2Y11P2Y purinoceptor 11, purinergic receptor P2Y, G-protein coupled, 11, purinergic receptor P2Y11 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.