Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human P2Y11/P2RY11 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9305N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9305N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9305N100 Supplier R&D Systems Supplier No. FAB9305N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

P2Y11/P2RY11 Monoclonal specifically detects P2Y11/P2RY11 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen P2Y11/P2RY11
Applications Flow Cytometry
Classification Monoclonal
Clone 505214
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q96G91
Gene Alias P2RY11, P2Y11P2Y purinoceptor 11, purinergic receptor P2Y, G-protein coupled, 11, purinergic receptor P2Y11
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human P2Y11/P2RY11 Accession # Q96G91
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5032
Test Specificity Stains human P2Y11/P2RY11 transfectants but not irrelevant transfectants in flow cytometry.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.