Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human p300 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3789
This item is not returnable.
View return policy
Description
KAT3B/p300 Polyclonal specifically detects KAT3B/p300 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.Specifications
KAT3B/p300 | |
Polyclonal | |
Unconjugated | |
Q09472 | |
EP300 | |
E. coli-derived recombinant human p300 His2283-His2414 Accession # Q09472 | |
100 μg | |
Primary | |
Detects human p300 in Western blots. | |
Human | |
IgG |
ChIP Assay, Immunocytochemistry, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-25 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 1-15 ug/mL | |
E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, EP300, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 | |
Goat | |
Affinity Purified | |
RUO | |
2033 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction