Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human p300 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3789
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3789 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3789 Supplier R&D Systems Supplier No. BAF3789
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

KAT3B/p300 Polyclonal specifically detects KAT3B/p300 in Human samples. It is validated for Western Blot.

Specifications

Antigen KAT3B/p300
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q09472
Gene Alias E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, EP300, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2
Gene Symbols EP300
Host Species Goat
Immunogen E. coli-derived recombinant human p300 His2283-His2414 Accession # Q09472
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2033
Test Specificity Detects human p300 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.