Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human p38 delta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1519
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1519 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1519 Supplier R&D Systems Supplier No. AF1519
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

p38 delta/SAPK4 Polyclonal specifically detects p38 delta/SAPK4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen p38 delta/SAPK4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q15759
Gene Alias EC 2.7.11, EC 2.7.11.24, MAP kinase 13, MAPK 13, MAPK13, MGC99536, mitogen-activated protein kinase 13, Mitogen-activated protein kinase p38 delta, p38delta, PRKM13, SAPK4MAP kinase p38 delta, Stress-activated protein kinase 4
Gene Symbols MAPK13
Host Species Rabbit
Immunogen E. coli-derived recombinant human p38 delta Accession # O15264
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5603
Test Specificity Detects endogenous human p38 delta . Mouse and rat p38 delta reactivity has not been demonstrated but is likely due to their high homology with human p38 delta . This antibody does not detect recombinant p38 alpha , p38 beta or p38 gamma in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.