Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human p62/SQSTM1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB80281SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB80281SP 25 μg
MAB80281100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB80281SP Supplier R&D Systems Supplier No. MAB80281SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

p62/SQSTM1 Monoclonal specifically detects p62/SQSTM1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Knockout Validated.

Specifications

Antigen p62/SQSTM1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, Immunoassay
Classification Monoclonal
Clone 2533B
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Immunocytochemistry 8-25 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q13501
Gene Alias A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, SQSTM1, Ubiquitin-binding protein p62, ZIP3
Host Species Rabbit
Immunogen E. coli-derived recombinant human p62/SQSTM1, Asp368-Leu440, Accession # Q13501
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8878
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.