Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human p63/TP73L Antibody, R&D Systems™



Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF1916SP
This item is not returnable.
View return policy
Description
p63/TP73L Polyclonal specifically detects p63/TP73L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
p63/TP73L | |
Polyclonal | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q9H3D4 | |
TP63 | |
E. coli-derived recombinant human p63/TP73L Met40-Cys339 Accession # Q9H3D4 | |
25 μg | |
Primary | |
Detects human p63/TP73L in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
amplified in squamous cell carcinoma, Chronic ulcerative stomatitis protein, CUSP, EEC3, EEC3B(p51B), Keratinocyte transcription factor KET, KET, KETRHS, LMS, OFC8, OFC8SHFM4TP73LNBP, p40, p51, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), SHFM4, TP53CP, TP53L, TP63, TP73L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like | |
Goat | |
Affinity Purified | |
RUO | |
8626 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction