Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human p63/TP73L Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF1916
This item is not returnable.
View return policy
Description
p63/TP73L Polyclonal specifically detects p63/TP73L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
p63/TP73L | |
Polyclonal | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
amplified in squamous cell carcinoma, Chronic ulcerative stomatitis protein, CUSP, EEC3, EEC3B(p51B), Keratinocyte transcription factor KET, KET, KETRHS, LMS, OFC8, OFC8SHFM4TP73LNBP, p40, p51, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), SHFM4, TP53CP, TP53L, TP63, TP73L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like | |
Goat | |
Affinity Purified | |
RUO | |
8626 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Q9H3D4 | |
TP63 | |
E. coli-derived recombinant human p63/TP73L Met40-Cys339 Accession # Q9H3D4 | |
100 μg | |
Primary | |
Detects human p63/TP73L in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction