Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human p63/TP73L Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1916
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1916 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1916 Supplier R&D Systems Supplier No. BAF1916
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

p63/TP73L Polyclonal specifically detects p63/TP73L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen p63/TP73L
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q9H3D4
Gene Alias amplified in squamous cell carcinoma, Chronic ulcerative stomatitis protein, CUSP, EEC3, EEC3B(p51B), Keratinocyte transcription factor KET, KET, KETRHS, LMS, OFC8, OFC8SHFM4TP73LNBP, p40, p51, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), SHFM4, TP53CP, TP53L, TP63, TP73L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like
Gene Symbols TP63
Host Species Goat
Immunogen E. coli-derived recombinant human p63/TP73L Met40-Cys339 Accession # Q9H3D4
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8626
Test Specificity Detects p63/TP73L in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.