Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human p70 S6 Kinase beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2987SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2987SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2987SP Supplier R&D Systems Supplier No. AF2987SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

p70 S6 Kinase beta/S6K2 Polyclonal specifically detects p70 S6 Kinase beta/S6K2 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen p70 S6 Kinase beta/S6K2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UBS0
Gene Alias EC 2.7.11, EC 2.7.11.1, KLS, p70 ribosomal S6 kinase beta, p70 S6 kinase beta, p70 S6KB, p70 S6K-beta, p70(S6K)-beta, p70-beta, P70-beta-1, P70-beta-2, p70-S6K 2, P70S6K2, p70S6Kb, ribosomal protein S6 kinase beta-2, ribosomal protein S6 kinase, 70kD, polypeptide 2,70 kDa ribosomal protein S6 kinase 2, ribosomal protein S6 kinase, 70kDa, polypeptide 2, RPS6KB2, S6K2, S6K-beta, S6K-beta2, S6K-beta-2, serine/threonine-protein kinase 14 beta, Serine/threonine-protein kinase 14B, SRK, STK14B, STK14BS6 kinase-related kinase
Gene Symbols RPS6KB2
Host Species Goat
Immunogen E. coli-derived recombinant human p70 S6 Kinase beta Ala2-Ser410 Accession # Q9UBS0
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6199
Test Specificity Detects human p70 S6 Kinase beta in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human p70 S6 Kinase is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.