Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PA2G4 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $410.00
Specifications
Antigen | PA2G4/EBP1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
PA2G4/EBP1 Polyclonal specifically detects PA2G4/EBP1 in Human samples. It is validated for Western Blot.Specifications
PA2G4/EBP1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Cell cycle protein p38-2G4 homolog, EBP1, ErbB-3 binding protein 1, ErbB3-binding protein 1, ErbB3-binding protein Ebp1, HG4-1, p38-2G4, proliferation-associated 2G4, 38kD, proliferation-associated 2G4, 38kDa, proliferation-associated protein 2G4 | |
PA2G4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9UQ80 | |
5036 | |
E. coli-derived recombinant human PA2G4 Ser2-Asp394 Accession # Q9UQ80 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human PA2G4 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title