Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human PACAP-27 ELISA Kit (Colorimetric)
SDP

Catalog No. NB398015
Click to view available options
Quantity:
1 Kit

Kit (Colorimetric)

Specifications

Assay Range 0.31 to 20 ng/mL
Assay Sensitivity 0.19 ng/mL
Detection Method ELISA
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Supernatant
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 116
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym ADCYAP1, adenylate cyclase activating polypeptide 1 (pituitary), PACAP, PACAP27, pituitary adenylate cyclase-activating polypeptide, Pituitary adenylate cyclase-activating polypeptide 27, prepro-PACAP
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.