Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PADI2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7257
This item is not returnable.
View return policy
Description
PADI2 Polyclonal specifically detects PADI2 in Human samples. It is validated for Immunohistochemistry.Specifications
PADI2 | |
Polyclonal | |
Unconjugated | |
Q9Y2J8 | |
PADI2 | |
E. coli-derived recombinant human PADI2 Met1-Ala115 Accession # Q9Y2J8 | |
100 μg | |
Primary | |
Detects human PADI2 in direct ELISAs. | |
Human | |
IgG |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
EC 3.5.3.15, KIAA0994PDI2PAD2, PAD2, PAD-H19, PDI2, peptidlyarginine deiminase type II, peptidyl arginine deiminase, type II, Peptidylarginine deiminase II, protein arginine deiminase, Protein-arginine deiminase type II, protein-arginine deiminase type-2 | |
Sheep | |
Affinity Purified | |
RUO | |
11240 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction