Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PADI2 Partial ORF (NP_003008, 1 a.a. - 108 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7099181
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. The type II enzyme is the most widely expressed family member. Known substrates for this enzyme include myelin basic protein in the central nervous system and vimentin in skeletal muscle and macrophages. This enzyme is thought to play a role in the onset and progression of neurodegenerative human disorders, including Alzheimer disease and multiple sclerosis, and it has also been implicated in glaucoma pathogenesis. This gene exists in a cluster with four other paralogous genes. [provided by RefSeq]

Sequence: MLRERTVRLQYGSRVEAVYVLGTYLWTDVYSAAPAGAQTFSLKHSEHVWVEVVRDGEAEEVATNGKQRWLLSPSTTLRVTMSQASTEASSDKVTVNYYDEEGSIPIDQ

Specifications

Accession Number NP_003008
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 11240
Molecular Weight (g/mol) 37.62kDa
Name PADI2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MLRERTVRLQYGSRVEAVYVLGTYLWTDVYSAAPAGAQTFSLKHSEHVWVEVVRDGEAEEVATNGKQRWLLSPSTTLRVTMSQASTEASSDKVTVNYYDEEGSIPIDQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA0994/PAD-H19/PAD2/PDI2
Common Name PADI2
Gene Symbol PADI2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.