Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PAG1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5285SP
This item is not returnable.
View return policy
Description
PAG1 Monoclonal specifically detects PAG1 in Human samples. It is validated for Western Blot, Simple Western.Specifications
PAG1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
Q9NWQ8 | |
PAG1 | |
E. coli-derived human PAG1 Ser38-Leu432 Accession # Q9NWQ8 | |
25 μg | |
Primary | |
Detects human PAG1 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
504004 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CBP, CBPMGC138364, Csk-binding protein, PAGFLJ37858, phosphoprotein associated with glycosphingolipid microdomains 1, phosphoprotein associated with glycosphingolipid-enriched microdomains 1, Transmembrane adapter protein PAG, Transmembrane phosphoprotein Cbp | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
55824 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction