Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Pancreatic Polypeptide/PP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6297
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6297 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6297 Supplier R&D Systems Supplier No. AF6297
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Pancreatic Polypeptide/PP Polyclonal specifically detects Pancreatic Polypeptide/PP in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Pancreatic Polypeptide/PP
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. P01298
Gene Alias INN, Obinepitide, pancreatic polypeptide (ppy), pancreatic polypeptide PNPPP, pancreatic polypeptide Y, pancreatic prohormone, PNP, PP, PPY
Gene Symbols PPY
Host Species Goat
Immunogen E. coli-derived recombinant human Pancreatic Polypeptide/PP Ala30-Leu95 Accession # P01298
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5539
Test Specificity Detects human Pancreatic Polypeptide/PP in direct ELISAs. In direct ELISAs, less than 15% cross-reactivity with recombinant mouse PPY is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.