Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Papilloma Virus 16 Mouse anti-Human papilloma virus, Clone: HPV16/1296, Abnova™

Catalog No. 89035127 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-127 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-127 Supplier Abnova Corporation Supplier No. MAB15107
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human papilloma virus 16.

  • High specificity
  • High reactivity

Specifications

Antigen Human Papilloma Virus 16
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone HPV16/1296
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human papilloma virus 16.
Dilution Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS.
Host Species Mouse
Immunogen Human papilloma virus type 16.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human Papillomavirus (HPV)
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.