Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Papilloma Virus 16 Mouse anti-Human papilloma virus, Clone: HPV16/1296, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB15106
Description
Mouse monoclonal antibody raised against human papilloma virus 16.
- High specificity
- High reactivity
Specifications
Human Papilloma Virus 16 | |
Monoclonal | |
Unconjugated | |
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
Protein A/G purification | |
RUO | |
Human Papillomavirus (HPV) | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
HPV16/1296 | |
Mouse monoclonal antibody raised against human papilloma virus 16. | |
In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
Human papilloma virus type 16. | |
100 μg | |
Primary | |
Store at 4°C. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction