Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human papilloma virus 16 Mouse, Clone: CAMVIR-1, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14712
Description
Mouse monoclonal antibody raised against human papilloma virus 16 (HPV-16).
- High specificity
- High reactivity
Specifications
| Human papilloma virus 16 | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| Reacts with a protein of 57kDa which identified as the L1 protein of HPV-16. It is the major capsid protein of HPV-16. The antibody reacts with HPV-16, 33 and 37. | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| CAMVIR-1 | |
| Mouse monoclonal antibody raised against human papilloma virus 16 (HPV-16). | |
| In 10 mM PBS. | |
| HPV-16, major capsid protein L1. | |
| 100 μg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction