Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Papilloma Virus 16 Mouse, Clone: HPV16L1/1058, Abnova™

Catalog No. 89034799 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-799 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-799 Supplier Abnova Corporation Supplier No. MAB14716
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human papilloma virus 16 (HPV-16).

  • High specificity
  • High reactivity

Specifications

Antigen Human Papilloma Virus 16
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HPV16L1/1058
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human papilloma virus 16 (HPV-16).
Dilution Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Immunoprecipitation (0.5-1 ug/500 ug protein Lysate) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determin
Formulation In 10 mM PBS.
Host Species Mouse
Immunogen HPV-16, major capsid protein L1.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with a protein of 57kDa which identified as the L1 protein of HPV-16. It is the major capsid protein of HPV-16. The antibody reacts with HPV-16, 33 and 37.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.