Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Human Papillomavirus (HPV) Verification Panel
Shop All Microbiologics Products

Click to view available options
Quantity:
20
Description
- Pellets are stable at room-temperature
- For in vitro diagnostic use
- Control includes DNA sequences for E6/E7 amplification of HPV 16, 18 and 45 types
- Does not contain human beta globin
- Currently not available for shipment to Canada
Specifications
Specifications
| Certifications/Compliance | IVD, CE Marked |
| Content And Storage | Room Temperature |
| Format | Helix Elite |
| Manufacturing Origin | United States |
| Product Type | Helix Elite Control Reference Panels and Standards |
| For Use With (Application) | In vitro diagnostics |
| Packaging Type | Plastic Box |
| Quantity | 20 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction