Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Papillomavirus type 18 L1-Capsids Antibody (IgG) ELISA Kit (Colorimetric)
SDP

Catalog No. NB601061KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB601061KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB601061KT Supplier Novus Biologicals™ Supplier No. NBP2601061KIT
Add to Cart
Add to Cart

High sensitivity and specificity for detection of human HPV18L1 antibody (IgG).

The Human Papillomavirus type 18 L1-Capsids Antibody (IgG) ELISA Kit (Colorimetric) measures human Papillomavirus type 18 L1-Capsids Antibody (IgG) in serum, plasma.

Specifications

Assay Range Qualitative
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Specificity For detection of human HPV18L1 antibody (IgG)
Target Species Human
For Use With (Application) ELISA
Kit Contents Adhesive Strip (For 96 wells), Coated assay plate, HRP-conjugate Diluent, HRP-conjugate (100 x concentrate), TMB Substrate, Wash Buffer (25 x concentrate), Sample Diluent, Positive Control, Negative Control, Instruction manual
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Cell Biology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target Papillomavirus type 18 L1-Capsids Antibody (IgG)
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.