Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Pappalysin-1/PAPP-A Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2487
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2487 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2487 Supplier R&D Systems Supplier No. BAF2487
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Pappalysin-1/PAPP-A Polyclonal specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot.

Specifications

Antigen Pappalysin-1/PAPP-A
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q13219
Gene Alias ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4ase, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA, PAPPA1, PAPP-A1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, Pappalysin1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1
Gene Symbols PAPPA
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Pappalysin-1/PAPP-A (R&D Systems, Catalog # 2487-ZN) Glu82-Asp1214 Accession # Q13219
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5069
Test Specificity Detects human Pappalysin-1/PAPP-A in Western blots. In this format, less than 5% cross-reactivity with recombinant human PAPP-A2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.