Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pappalysin-2/PAPP-A2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF1668SP
This item is not returnable.
View return policy
Description
Pappalysin-2/PAPP-A2 Polyclonal specifically detects Pappalysin-2/PAPP-A2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Pappalysin-2/PAPP-A2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.24.-, PAPPA2, PAPP-A2, PAPP-A2pappalysin-2, pappalysin 2, Pappalysin2, PAPPE, PAPP-E, PLAC3, placenta-specific 3, pregnancy-associated plasma preproprotein-A2, Pregnancy-associated plasma protein A2, Pregnancy-associated plasma protein E1 | |
Goat | |
Affinity Purified | |
RUO | |
60676 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q9BXP8 | |
PAPPA2 | |
Mouse myeloma cell line NS0-derived recombinant human Pappalysin-2/PAPP-A2 Ser234-Cys1396 Accession # Q9BXP8 | |
25 μg | |
Primary | |
Detects human Pappalysin-2/PAPP-A2 in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human PAPP-A is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction