Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PAR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3855
This item is not returnable.
View return policy
Description
PAR1/Thrombin Receptor Monoclonal specifically detects PAR1/Thrombin Receptor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
PAR1/Thrombin Receptor | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, CyTOF-ready | |
P25116 | |
F2R | |
E. coli-derived recombinant human PAR1 Arg27-Thr102, Ser375-Thr425 Accession # P25116 | |
100 μg | |
Primary | |
Detects human PAR1 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
731115 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CF2R, CF2RHTR, coagulation factor II (thrombin) receptor, Coagulation factor II receptor, F2R, HTR, PAR1, PAR1PAR-1protease-activated receptor 1, Protease-Activated Receptor 1, Thrombin receptor, ThrR, TRGPC, TRproteinase-activated receptor 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2149 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction