Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PAR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$176.44 - $461.02
Specifications
| Antigen | PAR1/Thrombin Receptor |
|---|---|
| Clone | 1028505 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB38551SP
![]() |
R&D Systems
MAB38551SP |
25 μg |
Each for $176.44
|
|
|||||
MAB38551100
![]() |
R&D Systems
MAB38551100 |
100 μg |
Each for $461.02
|
|
|||||
Description
PAR1/Thrombin Receptor Monoclonal specifically detects PAR1/Thrombin Receptor in Human samples. It is validated for Immunohistochemistry.Specifications
| PAR1/Thrombin Receptor | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| CF2R, CF2RHTR, coagulation factor II (thrombin) receptor, Coagulation factor II receptor, F2R, HTR, PAR1, PAR1PAR-1protease-activated receptor 1, Protease-Activated Receptor 1, Thrombin receptor, ThrR, TRGPC, TRproteinase-activated receptor 1 | |
| Human PAR1 synthetic peptide, Accession # P25116 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1028505 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P25116 | |
| 2149 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title