Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PAR2 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3949N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3949N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3949N100 Supplier R&D Systems Supplier No. FAB3949N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PAR2 Monoclonal specifically detects PAR2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PAR2
Applications Flow Cytometry
Classification Monoclonal
Clone 344222
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P55085
Gene Alias CFIIRL1, coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, F2RL1, GPCR11, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1
Gene Symbols F2RL1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human PAR2 Accession # P55085
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2150
Test Specificity Detects human PAR2. Stains human PAR2 transfectants but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.