Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PARD3/Par3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8030
This item is not returnable.
View return policy
Description
PARD3/Par3 Monoclonal specifically detects PARD3/Par3 in Human samples. It is validated for Immunocytochemistry.Specifications
| PARD3/Par3 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| Q8TEW0 | |
| PARD3 | |
| E. coli-derived recombinant human PARD3/Par3 Tyr451-Glu555 Accession # Q8TEW0 | |
| 100 μg | |
| Primary | |
| Detects human PARD3/Par3 in ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human PARD3b/Par3b is observed. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 833517 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ASIP, Baz, Bazooka, FLJ21015, PAR3, par-3 partitioning defective 3 homolog (C. elegans), PAR3A, PAR3C.elegans) homolog, SE2-5L16, SE2-5LT1, SE2-5T2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56288 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction