Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Parechovirus Type 1/3 VP1 Recombinant Rabbit Monoclonal Antibody (HL2427)
GREENER_CHOICE

Catalog No. FEMA550527
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550527 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550527 Supplier Invitrogen™ Supplier No. MA550527
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Human Parechovirus Type 1/3 VP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Human Parechovirus Type 1/3 VP1 is a gene target that encodes for a protein found in human parechoviruses (HPeV). HPeV is a common cause of mild to severe infections in infants and young children, and can also affect adults. The VP1 protein is located on the surface of the virus and is involved in the attachment and entry of the virus into host cells. Detection of this gene target can aid in the diagnosis and management of HPeV infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Parechovirus Type 1/3 VP1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2427
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Human PeV Type 1/3 VP1
Host Species Rabbit
Immunogen Full length of Human Parechovirus 3 VP1 recombinant protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.