Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Parkin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1438SP
This item is not returnable.
View return policy
Description
Parkin Monoclonal specifically detects Parkin in Human samples. It is validated for Western Blot.Specifications
Parkin | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O60260 | |
PARK2 | |
E. coli-derived recombinant human Parkin Met1-Val465 Accession # O60260 | |
25 μg | |
Primary | |
Detects human Parkin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Parkin-2 is observed. | |
Human | |
Purified |
Western Blot | |
323302 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AR-JP, E3 ubiquitin ligase, E3 ubiquitin-protein ligase parkin, EC 6.3.2.-, LPRS2, PARK2, parkin, parkin 2, Parkinson disease protein 2, Parkinson juvenile disease protein 2, parkinson protein 2, E3 ubiquitin protein ligase (parkin), PDJ, PDJjuvenile) 2, parkin, PRKN | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5071 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction