Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Parkin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB14382SP
This item is not returnable.
View return policy
Description
Parkin Monoclonal specifically detects Parkin in Human samples. It is validated for Immunohistochemistry.Specifications
| Parkin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AR-JP, E3 ubiquitin ligase, E3 ubiquitin-protein ligase parkin, EC 6.3.2.-, LPRS2, PARK2, parkin, parkin 2, Parkinson disease protein 2, Parkinson juvenile disease protein 2, parkinson protein 2, E3 ubiquitin protein ligase (parkin), PDJ, PDJjuvenile) 2, parkin, PRKN | |
| E. coli-derived recombinant human Parkin, Met1-Val437, Accession # NP_054642 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry | |
| 328110 | |
| Immunohistochemistry 5-25 ug/mL | |
| NP_054642 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5071 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction