Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Parvalbumin ELISA Kit (Colorimetric)
SDP

Catalog No. NB171843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171843 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171843 Supplier Novus Biologicals™ Supplier No. NBP318221
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 39-2500 pg/ml
Assay Sensitivity 17.2 pg/ml
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 5816
Gene Symbol PVALB
Quantity 1 Kit
Research Discipline Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.