Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Patched 2/PTCH2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4078SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4078SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4078SP Supplier R&D Systems Supplier No. MAB4078SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Patched 2/PTCH2 Monoclonal specifically detects Patched 2/PTCH2 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Patched 2/PTCH2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 403438
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y6C5
Gene Alias patched (Drosophila) homolog 2, patched 2, patched homolog 2, patched homolog 2 (Drosophila), protein patched homolog 2, ptc2, PTCH2
Gene Symbols PTCH2
Host Species Mouse
Immunogen E. coli-derived recombinant human Patched 2/PTCH2 Glu79-Glu392 Accession # Q9Y6C5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8643
Test Specificity Detects human Patched 2/PTCH2 in direct ELISA and Western Blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.