Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Patched 2/PTCH2 (First Extracellular Loop) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4078
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4078 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4078 Supplier R&D Systems Supplier No. AF4078
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Patched 2/PTCH2 Polyclonal specifically detects Patched 2/PTCH2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen Patched 2/PTCH2
Applications Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Gene Accession No. Q9Y6C5
Gene Alias patched (Drosophila) homolog 2, patched 2, patched homolog 2, patched homolog 2 (Drosophila), protein patched homolog 2, ptc2, PTCH2
Gene Symbols PTCH2
Host Species Goat
Immunogen E. coli-derived recombinant human Patched 2/PTCH2 first extracellular loop Glu79-Glu392 Accession # Q9Y6C5
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8643
Test Specificity Detects human Patched 2/PTCH2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse Patched is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.