Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PAUF/ZG16B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7777SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7777SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7777SP Supplier R&D Systems Supplier No. MAB7777SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PAUF/ZG16B Monoclonal specifically detects PAUF/ZG16B in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PAUF/ZG16B
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 817310
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96DA0
Gene Alias DKFZp762P2216, EC 3.1.2, EC 3.4.19.12, EECP, HRPE773, hTrabid, JCLN2, LOC124220, PRO1567, TRABIDubiquitin thioesterase ZRANB1, TRAF-binding protein domain, ZG16B, Zinc finger Ran-binding domain-containing protein 1, zinc finger, RAN-binding domain containing 1
Gene Symbols ZG16B
Host Species Mouse
Immunogen E. coli-derived recombinant human PAUF/ZG16B Gly53-Arg208 Accession # Q96DA0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124220
Test Specificity Detects human PAUF/ZG16B in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.