Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pax3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$172.76 - $489.78
Specifications
| Antigen | Pax3 |
|---|---|
| Clone | 274206 |
| Dilution | Immunohistochemistry 5-15 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11430SP
![]() |
R&D Systems
MAB11430SP |
25 μg |
Each for $172.76
|
|
|||||
MAB11430100
![]() |
R&D Systems
MAB11430100 |
100 μg |
Each for $489.78
|
|
|||||
Description
Pax3 Monoclonal antibody specifically detects Pax3 in Human samples. It is validated for ImmunohistochemistrySpecifications
| Pax3 | |
| Immunohistochemistry 5-15 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3 | |
| E. coli-derived recombinant human PAX-3, Met1-Leu215, Accession # P23760 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 274206 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P23760 | |
| 5077 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title