Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pax4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$223.93 - $579.52
Specifications
| Antigen | Pax4 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
Description
Pax4 Polyclonal specifically detects Pax4 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| Pax4 | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4 | |
| PAX4 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| NP_006184 | |
| 5078 | |
| E. coli-derived recombinant human Pax4 Met1-Glu343 Accession # NP_006184 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Pax4 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Pax3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title