Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Pax6 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC8150P100
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC8150P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC8150P100 Supplier R&D Systems Supplier No. IC8150P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Pax6 Polyclonal specifically detects Pax6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Pax6
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P26367
Gene Alias keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6
Host Species Sheep
Immunogen E. coli-derived recombinant human Pax6 Met1-Arg272 Accession # P26367
Purification Method Affinity Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 5080
Test Specificity Detects human Pax6 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Pax1, rhPax2, rhPax3, rhPax4, rhPax5, and rhPax7 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.