Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PCK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7316
This item is not returnable.
View return policy
Description
PCK1 Monoclonal specifically detects PCK1 in Human samples. It is validated for Western Blot.Specifications
| PCK1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P35558 | |
| PCK1 | |
| E. coli-derived recombinant human PCK1 Met1-Ile88 Accession # P35558 | |
| 100 μg | |
| Primary | |
| Detects human PCK1 in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot | |
| 789114 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 4.1.1.32, MGC22652, PEPCK1, PEPCKC, PEPCK-C, PEPCK-CPEP carboxykinase, phosphoenolpyruvate carboxykinase 1 (soluble), phosphoenolpyruvate carboxykinase, cytosolic, phosphoenolpyruvate carboxykinase, cytosolic [GTP], Phosphoenolpyruvate carboxylase, phosphopyruvate carboxylase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5105 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction