Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PD-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
$227.50 - $2070.00
Specifications
| Antigen | PD-1 |
|---|---|
| Clone | 192106 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1086SP
![]() |
R&D Systems
MAB1086SP |
25 μg |
Each for $227.50
|
|
|||||
MAB1086100
![]() |
R&D Systems
MAB1086100 |
100 μg |
Each for $561.50
|
|
|||||
MAB1086500
![]() |
R&D Systems
MAB1086500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for Western Blot.Specifications
| PD-1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| PDCD1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 192106 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q8IX89 | |
| 5133 | |
| Mouse myeloma cell line NS0-derived recombinant human PD-1 Leu25-Gln167 Accession # Q8IX89 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PD-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CD28, rhCTLA4, rhICOS, or recombinant mouse PD-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title