Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PD-1 Fc Chimera Protein

Catalog No. 1086PD01M
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When Recombinant Human PD-1 Fc Chimera is immobilized at 0.1 μg/mL (100 μL/well), Recombinant Human B7-H1/PD-L1 Fc Chimera (Catalog # 156-B7 ) binds with a typical ED50 of 0.15-0.75 μg/mL.

Order Info

Ships at ambient temperature.

Specifications

Accession Number Q15116.3
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5133
Molecular Weight (g/mol) M.W. (Observed): 60 - 70 kDa, under reducing conditions.
Quantity 1 mg
Source Mouse myeloma cell line, NS0-derived human PD-1 protein Human PD-1 (Leu25-Gln167) (C-terminus) Accession # Q15116.3 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Gene Symbol PDCD1
Biological Activity 0.15 to 0.75μg/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-1
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.