Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PD-1 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7115P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7115P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7115P Supplier R&D Systems Supplier No. FAB7115P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PD-1 Polyclonal specifically detects PD-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PD-1
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q8IX89
Gene Alias CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Gene Symbols PDCD1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human PD-1 Leu25-Gln167 Accession # Q8IX89
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5133
Test Specificity Detects human PD-1 in ELISAs and Western blots. In sandwich ELISAs, less than 2% cross-reactivity with recombinant mouse PD-1 and less than 0.2% cross-reactivity with recombinant human (rh) CD28, rhICOS, and rhCTLA-4 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.