Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PD-ECGF/Thymidine Phosphorylase Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1646
This item is not returnable.
View return policy
Description
PD-ECGF/Thymidine Phosphorylase Monoclonal specifically detects PD-ECGF/Thymidine Phosphorylase in Human samples. It is validated for Western Blot.Specifications
PD-ECGF/Thymidine Phosphorylase | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P19971 | |
TYMP | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human PD-ECGF/Thymidine Phosphorylase Ala11-Gln482 Accession # P19971 | |
500 μg | |
Primary | |
Detects human PD-ECGF/Thymidine Phosphorylase in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
27917 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dThdPase, EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase, TPase, TYMP | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
1890 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction